missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human alpha-2b Adrenergic Receptor (aa 198-247) Control Fragment Recombinant Protein

Product Code. 30198058
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30198058

Brand: Invitrogen™ RP103896

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (57%), Rat (57%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64258 (PA5-64258. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Alpha-2-adrenergic receptors are members of the G protein-coupled receptor superfamily. They include 3 highly homologous subtypes: alpha2A, alpha2B, and alpha2C. These receptors have a critical role in regulating neurotransmitter release from sympathetic nerves and from adrenergic neurons in the central nervous system. Alpha 2B adrenergic receptor subtype was observed to associate with eIF-2B, a guanine nucleotide exchange protein that functions in regulation of translation. A polymorphic variant of the alpha2B subtype, which lacks 3 glutamic acids from a glutamic acid repeat element, was identified to have decreased G protein-coupled receptor kinase-mediated phosphorylation and desensitization; this polymorphic form is also associated with reduced basal metabolic rate in obese subjects and may therefore contribute to the pathogenesis of obesity. Alpha 2B adrenergic receptor gene contains no introns in either its coding or untranslated sequences.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P18089
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 151
Name Human alpha-2 b Adrenergic Receptor (aa 198-247) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias [a]2 B; ADA2B; Adra2b; Adra-2 b; ADRA2B adrenergic, alpha-2 B-, receptor; ADRA2L1; ADRA2RL1; ADRARL1; Adrenergic alpha2B- receptor class III; adrenergic receptor, alpha 2 b; adrenergic, alpha-2 B-, receptor; Adrenergic, alpha2B-, receptor class III; adrenoceptor alpha 2 B; alpha-2 adrenergic receptor subtype C2; alpha-2-adrenergic receptor-like 1; alpha2B; alpha-2 B adrenergic receptor; Alpha-2 B adrenoceptor; alpha-2 B adrenoreceptor; ALPHA2BAR; alpha-2 BAR; alpha2-C2; FAME2; G-protein coupled receptor; subtype alpha2-C2
Common Name alpha-2 b Adrenergic Receptor
Gene Symbol ADRA2B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IAKRSNRRGPRAKGGPGQGESKQPRPDHGGALASAKLPALASVASAREVN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.