missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha 2-Macroglobulin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1938-SP
This item is not returnable.
View return policy
Description
alpha 2-Macroglobulin Polyclonal specifically detects alpha 2-Macroglobulin in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Immunocytochemistry.
Specifications
| alpha 2-Macroglobulin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| A2M | |
| Human plasma-derived alpha 2 -Macroglobulin | |
| 25 μg | |
| Primary | |
| Detects human alpha 2 -Macroglobulin in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human alpha 2 -Macroglobulin-like 1 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 1 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| A2M, alpha 2Macroglobulin, Alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5, CPAMD5DKFZp779B086, FWP007, S863-7 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction