missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha-1B Adrenergic R/ADRA1B Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC4730S-100UG
This item is not returnable.
View return policy
Description
alpha-1B Adrenergic R/ADRA1B Monoclonal specifically detects alpha-1B Adrenergic R/ADRA1B in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| alpha-1B Adrenergic R/ADRA1B | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| a-1B AdrenergicR, ADRA1, ADRA1B, adrenergic, alpha-1B-, receptor, alpha-1B adrenergic receptor, alpha1B AdrenergicR, Alpha-1B adrenoceptor, Alpha-1B adrenoreceptor, alpha-1B-adrenergic receptor, ALPHA1BAR | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human ADRA1B. Stains human ADRA1B transfectants but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 471802 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P35368 | |
| ADRA1B | |
| NS0 mouse myeloma cell line transfected with human ADRA1B Met1-Phe520 Accession # P35368 | |
| RUO | |
| 147 | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction