missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha 1-Acid Glycoprotein Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3694-SP
This item is not returnable.
View return policy
Description
alpha 1-Acid Glycoprotein Polyclonal specifically detects alpha 1-Acid Glycoprotein in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| alpha 1-Acid Glycoprotein | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ORM1 | |
| Human plasma-derived alpha 1-Acid Glycoprotein | |
| 25 μg | |
| Primary | |
| Detects human alpha 1-Acid Glycoprotein in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mose alpha 1-Acid Glycoprotein is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 5 ug/mL | |
| AGP, AGP 1, AGP1, AGP1alpha-1-acid glycoprotein 1, AGPA, AGP-A, alpha 1Acid Glycoprotein, OMD 1, ORM, ORM1, Orosomucoid, orosomucoid 1, orosomucoid-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5004 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction