missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Alkaline Phosphatase/ALPL Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
£359.60
Specifications
Antigen | Alkaline Phosphatase/ALPL |
---|---|
Clone | 388816 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Alkaline Phosphatase/ALPL Monoclonal specifically detects Alkaline Phosphatase/ALPL in Human samples. It is validated for Western Blot.Specifications
Alkaline Phosphatase/ALPL | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Human | |
P05186 | |
249 | |
E. coli-derived recombinant human Alkaline Phosphatase/ALPL Leu18-Ser502 Accession # P05186 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Alkaline Phosphatase/ALPL in direct ELISAs and Western blots. |
388816 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Akp2, Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALP, TNSALPMGC167935 | |
ALPL | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only