missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ALK-1 ELISA Kit

Product Code. 17814090
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17814090

Brand: Invitrogen™ EH17RBX10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human ALK-1 quantitates human ALK-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ALK-1.

Activin A receptor type II-like 1 (ACVRL1) is a type I cell-surface receptor for the TGF-beta superfamily of ligands. ACVRL1, similar to other type 1 receptors, has a conserved serine-threonine kinase subdomain, a glycine- and serine-rich region (called the GS domain) preceding the kinase domain, and a short C-terminal tail. Mutations in ACVRL1 are associated with hemorrhagic telangiectasia type 2. Mutations in ACVRL1 have been linked to Fibrodysplasia Ossificans Progressiva (FOP), a rare genetic disorder characterized by the formation of bone in muscles, tendons, and other connective tissues, as well as Epicanthus.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number activin A receptor like type 1; activin A receptor type II-like 1; activin A receptor type IL; activin A receptor, type II-like 1; activin A receptor, type II-like kinase 1; Activin receptor like kinase 1; activin receptor-like kinase 1; activin receptor-like kinase-1; ACVRL1; ACVRLK1; AI115505; AI427544; ALK1; ALK-1; HHT; HHT2; ORW2; R3; R-3; Serine/threonine-protein kinase receptor R3; SETHKIR; SKR3; TGF-B superfamily receptor type I; TSR-I
Assay Range 0.18 to 40 ng/mL
Assay Sensitivity 0.18 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 94
Gene Symbol ACVRL1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target ACVRL1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.