missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ALK-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£164.00 - £417.00
Specifications
| Antigen | ALK-1 |
|---|---|
| Clone | 117702 |
| Dilution | Blockade of Receptor-ligand Interaction 0.15-0.75 μg/mL |
| Applications | Blocking Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18731347
![]() |
R&D Systems
MAB3701-SP |
25 μg |
£164.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18722367
![]() |
R&D Systems
MAB3701-100 |
100 μg |
£417.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALK-1 Monoclonal specifically detects ALK-1 in Human samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
| ALK-1 | |
| Blockade of Receptor-ligand Interaction 0.15-0.75 μg/mL | |
| Monoclonal | |
| Lyophilized | |
| Human | |
| P37023 | |
| 94 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 117702 | |
| Blocking Assay | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRL1, ACVRLK1ORW2, ALK1, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I | |
| Mouse myeloma cell line NS0-derived recombinant human ALK-1, Asp22-Gln118, Accession # P37023 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human ALK-1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title