missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ALDH5A1 (aa 297-396) Control Fragment Recombinant Protein

Product Code. 30204732
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30204732

Brand: Invitrogen™ RP94683

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83102 (PA5-83102. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Aldh5A1 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldehyde dehydrogenase enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. Aldh5A1 is a mitochondrial NAD(+)-dependent succinic semialdehyde dehydrogenase. A deficiency of this enzyme, known as 4-hydroxybutyricaciduria, results in a disorder of the neurotransmitter 4-aminobutyric acid (GABA). Symptoms usually include static encephalopathy, associated with developmental delays, hypotonia, ataxia, speech defects, and seizures. At least two isoforms of Aldh5A1 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P51649
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 7915
Name Human ALDH5A1 (aa 297-396) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6330403E24Rik; Ahd1; Ahd-1; aldehyde dehydrogenase 1, mitochondrial; aldehyde dehydrogenase 5 family member A1; aldehyde dehydrogenase 5 family, member A1; aldehyde dehydrogenase 5 family, member A1 (succinate-semialdehyde dehydrogenase); aldehyde dehydrogenase family 5 member A1; aldehyde dehydrogenase family 5, subfamily A1; ALDH5A1; aldhehyde dehydrogenase family 5, subfamily A1; D630032B01Rik; mitochondrial succinate semialdehyde dehydrogenase; NAD(+)-dependent succinic semialdehyde dehydrogenase; OTTMUSG00000000613; SSADH; SSDH; Ssdh1; Succinate-semialdehyde dehydrogenase, mitochondrial; succinic semialdehyde dehydrogenase
Common Name ALDH5A1
Gene Symbol ALDH5A1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ANSVKRVSMELGGLAPFIVFDSANVDQAVAGAMASKFRNTGQTCVCSNQFLVQRGIHDAFVKAFAEAMKKNLRVGNGFEEGTTQGPLINEKAVEKVEKQV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.