missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ALDH3A2 (aa 365-462) Control Fragment Recombinant Protein

Product Code. 30201833
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201833

Brand: Invitrogen™ RP89972

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53232 (PA5-53232. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Aldh3A2 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldehyde dehydrogenase enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. Aldh3A2 catalyzes the oxidation of long-chain aliphatic aldehydes to fatty acids. Mutations in the Aldh3A2 gene cause Sjogren-Larrson syndrome, an inherited neurocutaneous disorder. Patients with this disorder display ichthyosis, mental retardation and spastic diplegia. The pathogenesis of the cutaneous and neurological symptoms is thought to result from abnormal lipid accumulation in the membranes of skin and brain, the formation of aldehyde Schiff base adducts with amine-containing lipids or proteins, or defective eicosanoid metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P51648
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 224
Name Human ALDH3A2 (aa 365-462) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Ahd3; Ahd-3; Ahd-3 r; Ahd3-r; AI194803; alcohol dehydrogenase family 3, subfamily A2; aldehyde dehydrogenase 10; Aldehyde dehydrogenase 3; aldehyde dehydrogenase 3 family member A2; aldehyde dehydrogenase 3 family, member A2; aldehyde dehydrogenase 3A2; aldehyde dehydrogenase 4; Aldehyde dehydrogenase family 3 member A2; aldehyde dehydrogenase family 3 member A2; fatty aldehyde dehydrogenase; aldehyde dehydrogenase family 3, subfamily A2; ALDH10; Aldh3; Aldh3a2; Aldh4; Aldh4-r; DKFZp686E23276; FALDH; fatty aldehyde dehydrogenase; Fatty aldehyde dehydrogenase-like protein; FLJ20851; microsomal aldehyde dehydrogenase; msALDH; QccE-15682; SLS
Common Name ALDH3A2
Gene Symbol ALDH3A2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SHNHKLIKRMIDETSSGGVTGNDVIMHFTLNSFPFGGVGSSGMGAYHGKHSFDTFSHQRPCLLKSLKREGANKLRYPPNSQSKVDWGKFFLLKRFNKE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.