missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ALDH1A1 ELISA Kit
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
Description
Human ALDH1A1 quantitates human ALDH1A1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ALDH1A1.
ALDH1A1 belongs to the aldehyde dehydrogenases family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. Two major liver isoforms of this enzyme, cytosolic and mitochondrial, can be distinguished by their electrophoretic mobilities, kinetic properties, and subcellular localizations. Most Caucasians have two major isozymes, while approximately 50% of Orientals have only the cytosolic isozyme, missing the mitochondrial isozyme. A remarkably higher frequency of acute alcohol intoxication among Orientals than among Caucasians could be related to the absence of the mitochondrial isozyme.
Specifications
Specifications
| Accession Number | acetaldehyde dehydrogenase 1; Ahd2; Ahd-2; alcohol dehydrogenase family 1, subfamily A1; alcohol dehydrogenase family 1, subfamily A2; ALDC; aldehyde dehydrogenase 1 family member A1; aldehyde dehydrogenase 1 family, member A1; aldehyde dehydrogenase 1, liver cytosolic (class 1); aldehyde dehydrogenase 1, soluble; aldehyde dehydrogenase 1, subfamily A1; aldehyde dehydrogenase 1A1; aldehyde dehydrogenase 2; aldehyde dehydrogenase family 1 member A1; aldehyde dehydrogenase family 1, member A1; aldehyde dehydrogenase family 1, subfamily A1; aldehyde dehydrogenase, cytosolic; aldehyde dehydrogenase, liver cytosolic; Aldh; ALDH class 1; Aldh1; ALDH11; ALDH1A1; Aldh1a2; Aldh2; ALDH-E1; ALHDII; E1; epididymis luminal protein 12; epididymis luminal protein 9; epididymis secretory sperm binding protein Li 53 e; HEL12; HEL-9; HEL-S-53 e; MGC2318; PUMB1; RALDH 1; ralDH1; Retinal dehydrogenase 1; retinaldehyde dehydrogenase 1 |
| Assay Range | 0.4 to 100 ng/mL |
| Assay Sensitivity | 0.4 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 216 |
| Gene Symbol | ALDH1A1 |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction