missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Aldehyde Oxidase 1/AOX1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5928
This item is not returnable.
View return policy
Description
Aldehyde Oxidase 1/AOX1 Polyclonal specifically detects Aldehyde Oxidase 1/AOX1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| Aldehyde Oxidase 1/AOX1 | |
| Polyclonal | |
| Unconjugated | |
| Q06278 | |
| AOX1 | |
| E. coli-derived recombinant human Aldehyde Oxidase 1/AOX1 Asn302-Ser531 Accession # Q06278 | |
| 100 μg | |
| Primary | |
| Detects human Aldehyde Oxidase 1/AOX1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 3-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| aldehyde oxidase, aldehyde oxidase 1, AOAOH1, AOX1, EC 1.2.3.1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 316 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction