missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ALCAM/CD166 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB656-500
This item is not returnable.
View return policy
Description
ALCAM/CD166 Monoclonal specifically detects ALCAM/CD166 in Human samples. It is validated for Western Blot, Adhesion Blockade.
Specifications
| ALCAM/CD166 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 214 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Adhesion Blockade | |
| 105901 | |
| Western Blot 2 ug/mL, Adhesion Blockade | |
| Q13740 | |
| ALCAM | |
| Mouse myeloma cell line NS0-derived recombinant human ALCAM/CD166 Trp28-Ala526 Accession # Q13740 | |
| 500 μg | |
| Primary | |
| Detects human ALCAM/CD166 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) BCAM, recombinant mouse OCAM, rhMCAM, or rhEpCAM is observed. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction