missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Akt2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB23152-SP
This item is not returnable.
View return policy
Description
AKT2 Monoclonal specifically detects AKT2 in Human samples. It is validated for Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| AKT2 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P31751 | |
| AKT2 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Akt2 Asn2-Glu481 Accession # P31751 | |
| 25 μg | |
| Primary | |
| Detects human Akt2. Using direct ELISA, this antibody does not detect recombinant human (rh) Akt1 or rhAkt3. | |
| Human | |
| Purified |
| Immunohistochemistry, Flow Cytometry, CyTOF | |
| 302501 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 208 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction