missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AKAP13 (aa 2358-2463) Control Fragment Recombinant Protein

Product Code. 30200575
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200575

Brand: Invitrogen™ RP102442

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54078 (PA5-54078. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms containing c-terminal dbl oncogene homology (DH) and pleckstrin homology (PH) domains. The DH domain is associated with guanine nucleotide exchange activation for the Rho/Rac family of small GTP binding proteins, resulting in the conversion of the inactive GTPase to the active form capable of transducing signals. The PH domain has multiple functions. Therefore, these isoforms function as scaffolding proteins to coordinate a Rho signaling pathway, function as protein kinase A-anchoring proteins and, in addition, enhance ligand-dependent activity of estrogen receptors alpha and beta.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q12802
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 11214
Name Human AKAP13 (aa 2358-2463) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1700026G02Rik; 5730522G15Rik; 5830460E08Rik; A kinase (PRKA) anchor protein 13; AKAP13; AKAP-13; AKAP-Lbc; A-kinase anchor protein 13; A-kinase anchoring protein; A-kinase anchoring protein 13; ARHGEF13; Breast cancer nuclear receptor-binding auxiliary protein; Brx; c-lbc; guanine nucleotide exchange factor Lbc; HA-3; Ht31; Human thyroid-anchoring protein 31; LBC; LBC oncogene; lymphoid blast crisis oncogene; Non-oncogenic Rho GTPase-specific GTP exchange factor; p47; PRKA13; Protein kinase A-anchoring protein 13; PROTO-LB; PROTO-LBC; type II cAMP-dependent protein kinase anchoring protein Ht31
Common Name AKAP13
Gene Symbol AKAP13
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ARELKEQLHQKDQKILLLLEEKEMIFRDMAECSTPLPEDCSPTHSPRVLFRSNTEEALKGGPLMKSAINEVEILQGLVSGNLGGTLGPTVSSPIEQDVVGPVSLPR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.