missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AKAP1 (aa 223-363) Control Fragment Recombinant Protein

Product Code. 30206068
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30206068

Brand: Invitrogen™ RP90162

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (35%), Rat (35%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes an enzyme which reversibly catalyzes the phosphorolysis of purine nucleosides. The enzyme is trimeric, containing three identical subunits. Mutations which result in nucleoside phosphorylase deficiency result in defective T-cell (cell-mediated) immunity but can also affect B-cell immunity and antibody responses. Neurologic disorders may also be apparent in patients with immune defects. A known polymorphism at aa position 51 that does not affect enzyme activity has been described. A pseudogene has been identified on chromosome 2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92667
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8165
Name Human AKAP1 (aa 223-363) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A kinase (PRKA) anchor protein 1; a kinase anchor protein 1, mitochondrial; Akap; AKAP 121; AKAP 149; Akap1; AKAP121; AKAP149; AKAP84; A-kinase anchor protein 1, mitochondrial; A-kinase anchor protein 121 kDa; A-kinase anchor protein 149 kDa; A-kinase anchoring protein 1; C76494; C81186; DAKAP1; D-AKAP1; D-AKAP-1; Dual specificity A-kinase-anchoring protein 1; dual-specificity A-kinase anchoring protein 1; PPP1R43; PRKA1; protein kinase A anchoring protein 1; protein kinase A1; protein kinase A-anchoring protein 1; protein phosphatase 1, regulatory subunit 43; S-AKAP; SAKAP84; S-AKAP84; spermatid A-kinase; Spermatid A-kinase anchor protein; spermatid A-kinase anchor protein 84; TDRD17; testicular secretory protein Li 5; tudor domain containing 17
Common Name AKAP1
Gene Symbol AKAP1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EHVLELENSKGPSLASLEGEEDKGKSSSSQVVGPVQEEEYVAEKLPSRFIESAHTELAKDDAAPAPPVADAKAQDRGVEGELGNEESLDRNEEGLDRNEEGLDRNEESLDRNEEGLDRNEEIKRAAFQIISQVISEATEQV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.