Learn More
ACROBiosystems Human AIMP1 / EMAP2 / SCYE1 Protein, His Tag
Description
Aminoacyl tRNA synthase complex-interacting multifunctional protein 1 (AIMP1) is also known as multisynthase complex auxiliary component p43 and endothelial monocyte-activating polypeptide II (EMAP-II). AIMP1 is a cytokine that may be induced by apoptosis and is also released from professional antigen-presenting cells such as dendritic cells. The release of AIMP1 renders the tumor-associated vasculature sensitive to tumor necrosis factor. Furthermore, AIMP1 binds tRNA and stimulates the catalytic activity of cytoplasmic arginyl-tRNA synthase. This protein possesses inflammatory cytokine activity. Also, AIMP1 negatively regulates TGF-beta signaling through stabilization of SMURF2 by binding to SMURF2 and inhibiting its SMAD7-mediated degradation.
Specifications
Specifications
| Accession Number | AAH14051.1 |
| Molecular Weight (g/mol) | 35.2 kDa |
| Quantity | 100 μg |
| Source | Human AIMP1, His Tag (EM2-H5142) is expressed from E.coli cells. It contains AA Ala 2 - Lys 312 (Accession # Q12904-1). |
| Immunogen | Ala 2 - Lys 312 |
| Storage Requirements | -20°C |
| Gene Alias | AIMP1,EMAP2,EMAPII,HLD3,SCYE1,p43,EMAP-2,EMAP-II,AIMP-1 |
| Common Name | AIMP1 |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.