missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human AIM Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
AIM Synthetic Peptide, PEP-0362, from Invitrogen.
CD5L, also known as CD5 antigen-like, is a protein that is expressed on the surface of various immune cells, including T cells, B cells, and natural killer cells. It is involved in the regulation of immune responses and is being studied as a potential therapeutic target in various diseases, including autoimmune disorders, cancer, and infectious diseases. CD5L is known to modulate T cell activation and cytokine production, and has been shown to have anti-inflammatory effects.
Specifications
Specifications
| Gene ID (Entrez) | 922 |
| Species | Human |
| Purification Method | purified |
| Accession Number | O43866 |
| Common Name | Human AIM |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction