missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AICL/CLEC-2B Alexa Fluor™ 405-conjugated Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9059V-100UG
This item is not returnable.
View return policy
Description
AICL/CLEC-2B Monoclonal antibody specifically detects AICL/CLEC-2B in Human samples. It is validated for Flow Cytometry
Specifications
| AICL/CLEC-2B | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Q92478 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9976 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 1064935 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Activation-induced C-type lectin, AICLC-type lectin superfamily member 2, CLEC2B, CLEC-2B, CLECSF2, CLECSF2IFN-alpha-2b-inducing-related protein 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 2 (activation-induced), C-type lectin domain family 2 member B, C-type lectin domain family 2, member B, HP10085, IFN-alpha2b-inducing related protein 1, IFNRG1 | |
| Chinese Hamster Ovary cell line, CHO-derived human AICL/CLEC-2B, Lys26-His149, Accession # Q92478 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction