missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AgRP/ART Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF704
This item is not returnable.
View return policy
Description
AgRP/ART Polyclonal specifically detects AgRP/ART in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| AgRP/ART | |
| Polyclonal | |
| Unconjugated | |
| O00253 | |
| AGRP | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human AgRP/ART Ala21-Thr132 Accession # O00253 | |
| 100 μg | |
| Primary | |
| Detects human AgRP/ART in direct ELISAs and Western blots. In Western blots, approximately 40% cross-reactivity with recombinant mouse AgRP/ART is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| agouti related protein homolog (mouse), agouti-related protein, Agrt, AGRTMGC118963, ART, ARTagouti (mouse) related protein, ASIP2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 181 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction