missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Agrin Recombinant Protein, PeproTech®

Product Code. 17904192
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
17904192 50 μg 50µg
17974352 500 μg 500µg
17984182 250 μg 250µg
17984342 1 mg 1mg
17994182 10 μg 10µg
17904172 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17904192 Leverantör Gibco™ Leverantörsnummer 1600850UG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

Recombinant Human Agrin is 111.9 kDa protein containing 1073 amino acids, corresponding to the 110 kDa N-terminal subunit of the full length protein. As a result of glycosylation, Recombinant Human Agrin migrates with an apparent molecular mass of approximately 150-160 kDa by SDS-PAGE gel, under reducing and non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Agrin is a molecule that resides in the basal lamina of muscle cells and directs key events in post synaptic differentiation. Most notably, Agrin is responsible for the clustering of acetylcholine receptors (AChRs) on the cell surface and their localization to the neuromuscular junction. Several Agrin variants have been identified which arise from alternative mRNA splicings. Agrin splice forms having inserts at two sites in the carboxy terminus designated 'y' and 'z' display a high affinity for AChRs, while splice forms lacking these inserts associate with AChRs weakly. Muscle alpha-dystroglycan has been postulated to be the receptor for the clustering activity of agrin; however, this is a point of contention. Tyrosine phosphorylation has been implicated as a required early step in AChR aggregation. Interestingly, a unique receptor tyrosine kinase, designated MuSK, has been discovered that interacts with Agrin and is specifically localized to developing muscle.
TRUSTED_SUSTAINABILITY

Specifikationer

Accession Number O00468
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 375790
Molecular Weight (g/mol) 111.9 kDa
Name Human Agrin
Quantity 50 μg
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AGR; Agrin; Agrin C-terminal 110 kDa subunit; Agrin C-terminal 22 kDa fragment; Agrin C-terminal 90 kDa fragment; Agrin N-terminal 110 kDa subunit; agrin proteoglycan; Agrn; C22; C90; CMS8; CMSPPD; nmf380; RP23-139J18.2
Common Name Agrin
Biological Activity Determined by its ability to bind U-87 mg cells in a functional ELISA.
Conjugate Unconjugated
Recombinant Recombinant
Sequence TCPERALERR EEEANVVLTG TVEEILNVDP VQHTYSCKVR VWRYLKGKDL VARESLLDGG NKVVISGFGD PLICDNQVST GDTRIFFVNP APPYLWPAHK NELMLNSSLM RITLRNLEEV EFCVEDKPGT HFTPVPPTPP DACRGMLCGF GAVCEPNAEG PGRASCVCKK SPCPSVVAPV CGSDASTYSN ECELQRAQCS QQRRIRLLSR GPCGSRDPCS NVTCSFGSTC ARSADGLTAS CLCPATCRGA PEGTVCGSDG ADYPGECQLL RRACARQENV FKKFDGPCDP CQGALPDPSR SCRVNPRTRR PEMLLRPESC PARQAPVCGD DGVTYENDCV MGRSGAARGL LLQKVRSGQC QGRDQCPEPC RFNAVCLSRR GRPRCSCDRV TCDGAYRPVC AQDGRTYDSD CWRQQAECRQ QRAIPSKHQG PCDQAPSPCL GVQCAFGATC AVKNGQAACE CLQACSSLYD PVCGSDGVTY GSACELEATA CTLGREIQVA RKGPCDRCGQ CRFGALCEAE TGRCVCPSEC VALAQPVCGS DGHTYPSECM LHVHACTHQI SLHVASAGPC ETCGDAVCAF GAVCSAGQCV CPRCEHPPPG PVCGSDGVTY GSACELREAA CLQQTQIEEA RAGPCEQAEC GSGGSGSGED GDCEQELCRQ RGGIWDEDSE DGPCVCDFSC QSVPGSPVCG SDGVTYSTEC ELKKARCESQ RGLYVAAQGA CRGPTFAPLP PVAPLHCAQT PYGCCQDNIT AARGVGLAGC PSACQCNPHG SYGGTCDPAT GQCSCRPGVG GLRCDRCEPG FWNFRGIVTD GRSGCTPCSC DPQGAVRDDC EQMTGLCSCK PGVAGPKCGQ CPDGRALGPA GCEADASAPA TCAEMRCEFG ARCVEESGSA HCVCPMLTCP EANATKVCGS DGVTYGNECQ LKTIACRQGL QISIQSLGPC QEAVAPSTHP TSASVTVTTP GLLLSQALPA PPGALPLAPS STAHSQTTPP PSSRPRTTAS VPRTTVWPVL TVPPTAPSPA PSLVASAFGE SGSTDGSSDE ELSGDQEASG GGSGGLEPLE GSSVATPGPP VER
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.