missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AGR2 Synthetic Peptide

Product Code. 15225489
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15225489 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15225489 Supplier Invitrogen™ Supplier No. PEP1560
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic Peptide

This peptide corresponds to 17 amino acid peptide near the amino terminus of human AGR2. PEP-1560 can be used as a blocking peptide with polyclonal antibody PA5-34517. For use as a blocking peptide in Western blot, incubate the peptide with equal volume of antibody for 30 min at 37°C.

AGR2 (anterior gradient protein 2) and AGR3 are members of the anterior gradient homolog family. They are secreted cytoplasmic proteins which are involved in metastasis induction and p53 tumour supressor inhibition. They may serve as molecular markers and potential therapeutic targets for hormone-responsive breast tumours. AGR2 is ubiquitously expressed with upregulated expression in prostate cancer, breast cancer, lung cancer, renal carcinomas and endometrial carcinomas. AGR2 expression is positively correlated with that of the estrogen receptor (ER) and is negatively correlated with that of the epidermal growth factor receptor (EGFR).
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 10551
Species Human
Purification Method purified
Accession Number O95994
Common Name AGR2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias AG2; AG-2; AGR2; agr2 protein; Agr2h; AII; AII type II arginase; anterior gradient 2; anterior gradient 2 homolog; anterior gradient 2 homologue; anterior gradient 2, protein disulphide isomerase family member; anterior gradient homolog 2; anterior gradient protein 2 homolog; ARG2; arginase 2; arginase II; arginase type II; arginase, type II; arginase-2, mitochondrial; AU022422; epididymis secretory protein Li 116; Gob4; Gob-4; HAG-2; HEL-S-116; HPC8; kidney arginase; kidney-type arginase; L-arginine amidinohydrolase; L-arginine ureahydrolase; mAG-2; nonhepatic arginase; Non-hepatic arginase; PDIA17; protein disulfide isomerase family A, member 17; protein Gob-4; secreted cement gland homolog; secreted cement gland protein XAG-2 homolog; Type II arginase; UNQ515/PRO1030; XAG-2
Gene Symbol Agr2
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.