missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Aggrecan Antibody, R&D Systems™

Mouse Monoclonal Antibody
£181.00 - £560.00
Specifications
| Antigen | Aggrecan |
|---|---|
| Clone | 179503 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18782467
![]() |
R&D Systems
MAB11304-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18721847
![]() |
R&D Systems
MAB11304-100 |
100 μg |
£560.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aggrecan Monoclonal antibody specifically detects Aggrecan in Human samples. It is validated for ELISASpecifications
| Aggrecan | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| ACAN, AGC1, AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16, MSK16AGCAN, SEDK | |
| Mouse myeloma cell line NS0-derived recombinant human Aggrecan, Val20-Gly675, Accession # NP_037359 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 179503 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_037359 | |
| 176 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title