missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AFP native Protein from human urine, Iodination Grade

Product Code. 16028196
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 16028196

Brand: Abnova™ P5345.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Conjugation, SDS-PAGE

This gene encodes alpha-fetoprotein, a major plasma protein produced by the yolk sac and the liver during fetal life. Alpha-fetoprotein expression in adults is often associated with hepatoma or teratoma. However, hereditary persistance of alpha-fetoprotein may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and the alpha-fetoprotein and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. Alpha-fetoprotein is found in monomeric as well as dimeric and trimeric forms, and binds copper, nickel, fatty acids and bilirubin. The level of alpha-fetoprotein in amniotic fluid is used to measure renal loss of protein to screen for spina bifida and anencephaly. [provided by RefSeq]

Specifications

For Use With (Application) Antibody Production, Conjugation, SDS-PAGE
Formulation In 0.1M sodium phosphate buffer, 0.2M NaCl, pH 7.4 (0.09% sodium azide)
Gene ID (Entrez) 174
Name AFP (Human) Native Protein
Preparation Method Native protein purified from human cord serum
Quantity 100 μg
Storage Requirements Store at 4°C on dry atmosphere or store at -20°C for long term storage. After reconstitution, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FETA/HPAFP
Common Name AFP
Gene Symbol AFP
Species Human
Protein Tag None
Expression System Native protein purified from human cord serum
Form Liquid
Purity or Quality Grade ≥98% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.