missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AFF1 (aa 1023-1086) Control Fragment Recombinant Protein

Product Code. 30198557
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30198557

Brand: Invitrogen™ RP107722

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (73%), Rat (73%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67082 (PA5-67082. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Proto-oncogene AF-4 (or FEL) is a product of a chromosomal aberration of the human gene AFF1, which is associated with acute leukemias. The fusion of AF-4 on chromosome band 4q21 with the mixed lineage leukemia (MLL or HRX) gene on 11q23 results in a MLL-AF-4 chimeric transcription factor in which AF-4 contributes transcriptional effector properties and requires cell-specific accessory factors. MLL is involved in several chromosomal translocations associated with acute myeloid and lymphoid leukemia. The MLL-AF-4 fusion protein is expressed in all normal hematopoietic cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P51825
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4299
Name Human AFF1 (aa 1023-1086) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 9630032B01Rik; AF4; AF4/FMR2 family member 1; AF4/FMR2 family, member 1; AFF1; ALL1-fused gene from chromosome 4 protein; AW319193; FEL; homolog of human MLLT2 unidentified; MLLT2; Mllt2h; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 2; myeloid/lymphoid or mixed-lineage leukemia translocated to 2 homolog; PBM1; pre-B-cell monocytic leukemia partner 1; protein AF-4; Protein FEL; Proto-oncogene AF4; Rob; robotic; translocated to, 2
Common Name AFF1
Gene Symbol AFF1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VDLIKFIMSLKSFSDATAPTQEKIFAVLCMRCQSILNMAMFRCKKDIAIKYSRTLNKHFESSSK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.