missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADP-Sugar Pyrophosphatase/NUDT5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6414-SP
This item is not returnable.
View return policy
Description
ADP-Sugar Pyrophosphatase/NUDT5 Monoclonal specifically detects ADP-Sugar Pyrophosphatase/NUDT5 in Human samples. It is validated for Western Blot.
Specifications
| ADP-Sugar Pyrophosphatase/NUDT5 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL | |
| Q9UKK9 | |
| NUDT5 | |
| E. coli-derived recombinant human ADP-Sugar Pyrophosphatase/NUDT5 Glu2-Phe219 Accession # Q9UKK9 | |
| 25 μg | |
| Primary | |
| Detects human ADP-Sugar Pyrophosphatase/NUDT5 in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot | |
| 739044 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.6.1.-, EC 3.6.1.13, hYSAH1, nucleoside diphosphate linked moiety X-type motif 5, Nucleoside diphosphate-linked moiety X motif 5, nudix (nucleoside diphosphate linked moiety X)-type motif 5, Nudix motif 5, YSA1, YSA1H, YSA1HADP-sugar pyrophosphatase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11164 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction