missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Adiponutrin/PNPLA3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £506.00
Specifications
| Antigen | Adiponutrin/PNPLA3 |
|---|---|
| Clone | 847516 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18795975
![]() |
R&D Systems
MAB5208-100 |
100 μg |
£506.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18776225
![]() |
R&D Systems
MAB5208-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Adiponutrin/PNPLA3 Monoclonal antibody specifically detects Adiponutrin/PNPLA3 in Human samples. It is validated for ImmunohistochemistrySpecifications
| Adiponutrin/PNPLA3 | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Acylglycerol O-acyltransferase, adiponutrin, ADPN, ADPNdJ796I17.1, C22orf20, Calcium-independent phospholipase A2-epsilon, chromosome 22 open reading frame 20, EC 2.3.1.-, EC 2.7.7.56, EC 3.1.1.3, EC 4.2.3.4, FLJ22012, iPLA(2)epsilon, iPLA2-epsilon, patatin-like phospholipase domain containing 3, patatin-like phospholipase domain-containing protein 3, PNPLA3 | |
| E. coli -derived recombinant human Adiponutrin/PNPLA3, Arg160-Arg349, Accession # Q9NST1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 847516 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9NST1 | |
| 80339 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title