missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Adiponectin Recombinant Protein, PeproTech®

Product Code. 17804593
Click to view available options
Quantity:
100 μg
2 x 500 μg
25 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
250µg
25µg
500µg
5µg
Pack of 2
This item is not returnable. View return policy

Product Code. 17804593

Brand: Gibco™ 45024100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

450-24-1MG will be provided as 2 x 500 μg (450-24-500UG). Recombinant Human Adiponectin is a multimeric glycoprotein containing amino acids Glu-19 to Asn-244 of the adiponectin precursor protein fused to an N-terminal histidine tag. Monomeric glycosylated adiponectin migrates at an apparent molecular weight of approximately 35.0 kDa by SDS PAGE analysis under reducing conditions. The calculated molecular weight of Recombinant Human Adiponectin is 25.9 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Adiponectin is a protein hormone secreted exclusively by adipocytes that regulates the metabolism of lipids and glucose. Adiponectin has been shown to play a role in various physiological processes including homeostasis and obesity. Decreased levels of adiponectin are associated with obese individuals as well as insulin resistance and hyperinsulinemia (type 2 diabetes). In mice adiponectin has been shown to improve insulin signaling, thus ameliorating insulin resistance. Through activation of AMP kinase and PPAR-alpha, adiponectin can increase fatty acid transport, oxidation, and dissipation in skeletal muscle, leading to a reduction of intramyocellular lipid levels and thus improved insulin signaling.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15848
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 9370
Molecular Weight (g/mol) 25.9 kDa
Name Human Adiponectin
Quantity 100 μg
Source Insect cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 30 kDa adipocyte complement-related protein; 30 kDa; Acdc; Acrp30; Ad; adipo; adipocyte complement related protein; adipocyte complement related protein of 30 kDa; adipocyte complement-related 30 kDa protein; adipocyte complement-related protein of 30 kDa; adipocyte, C1Q and collagen domain containing; adipocyte, C1q and collagen domain-containing protein; Adipocyte-specific protein AdipoQ; adiponectin; adiponectin, C1Q and collagen domain containing; Adipoq; Adipose most abundant gene transcript 1 protein; adipose specific collagen-like factor; ADIPQTL1; ADPIOQ; ADPN; Apm1; APM-1; APN; GBP28; Gelatin-binding protein; gelatin-binding protein 28
Common Name Adiponectin
Gene Symbol ADIPOQ
Biological Activity Determined by a cytotoxic assay using M1 cells. The ED50 for this effect is 3.0-6.0 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence RGHHHHHHHH ETTTQGPGVL LPLPKGACTG WMAGIPGHPG HNGAPGRDGR DGTPGEKGEK GDPGLIGPKG DIGETGVPGA EGPRGFPGIQ GRKGEPGEGA YVYRSAFSVG LETYVTIPNM PIRFTKIFYN QQNHYDGSTG KFHCNIPGLY YFAYHITVYM KDVKVSLFKK DKAMLFTYDQ YQENNVDQAS GSVLLHLEVG DQVWLQVYGE GERNGLYADN DNDSTFTGFL LYHDTN
Content And Storage -20°C
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 97% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.