missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Adiponectin/Acrp30 ELISA Kit (Colorimetric)

Product Code. 18674331 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18674331 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18674331 Supplier Novus Biologicals™ Supplier No. KA0017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Assay Range 1 to 100 ng/mL
Assay Sensitivity 0.47 ng/mL (serum, plasma), 0.156 ng/mL (urine, CSF)
Product Type ELISA Kit (Colorimetric)
Sample Type Breast Milk, Cerebrospinal Fluid, Plasma (EDTA, Citrate, Heparin), Serum, Urine
For Use With (Application) ELISA, Quantification
Gene ID (Entrez) 9370
Gene Symbol ADIPOQ
Quantity 1 Kit
Research Discipline Diabetes Research, Lipid and Metabolism
Storage Requirements Store at 4°C.
Synonym ACDC, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, APM1APM-1, C1q and collagen domain-containing protein, GBP28apM1, Gelatin-binding protein
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.