missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Adenylate Cyclase 8 (aa 610-695) Control Fragment Recombinant Protein

Product Code. 30204343
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30204343

Brand: Invitrogen™ RP93838

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55041 (PA5-55041. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Adenylate cyclase is a membrane bound enzyme that catalyses the formation of cyclic AMP from ATP. The enzymatic activity is under the control of several hormones, and different polypeptides participate in the transduction of the signal from the receptor to the catalytic moiety. Stimulatory or inhibitory receptors (Rs and Ri) interact with G proteins (Gs and Gi) that exhibit GTPase activity and they modulate the activity of the catalytic subunit of the adenylyl cyclase.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P40145
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 114
Name Human Adenylate Cyclase 8 (aa 610-695) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AC8; ADCY3; ADCY8; adenylate cyclase 8; adenylate cyclase 8 (brain); adenylate cyclase type 8; Adenylate cyclase type VIII; adenylyl cyclase 8; adenylyl cyclase-8, brain; ATP pyrophosphate-lyase 8; AW060868; ca(2+)/calmodulin-activated adenylyl cyclase; epididymis secretory sperm binding protein Li 172 mP; HBAC1; HEL-S-172 mP; type VIII adenylyl cyclase
Common Name Adenylate Cyclase 8
Gene Symbol Adcy8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DRRNSGATFTEGSWSPELPFDNIVGKQNTLAALTRNSINLLPNHLAQALHVQSGPEEINKRIEHTIDLRSGDKLRREHIKPFSLMF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.