missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Adenylate Cyclase 4 (aa 491-582) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61717 (PA5-61717. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a member of the family of adenylate cyclases, which are membrane-associated enzymes that catalyze the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). Mouse studies show that adenylate cyclase 4, along with adenylate cyclases 2 and 3, is expressed in olfactory cilia, suggesting that several different adenylate cyclases may couple to olfactory receptors and that there may be multiple receptor-mediated mechanisms for the generation of cAMP signals.
Specifications
Specifications
| Accession Number | Q8NFM4 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 196883 |
| Name | Human Adenylate Cyclase 4 (aa 491-582) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | A cyclase IV; AC4; Adcy4; adenylate cyclase 4; adenylate cyclase type 4; Adenylate cyclase type IV; adenylyl cyclase 4; ATP pyrophosphate-lyase 4; KIAA4004; mKIAA4004 |
| Common Name | Adenylate Cyclase 4 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction