missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Adenosine A2b R Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB104442T-100UG
This item is not returnable.
View return policy
Description
Adenosine A2b R Monoclonal specifically detects Adenosine A2b R in Human samples. It is validated for Flow Cytometry.
Specifications
| Adenosine A2b R | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| adenosine A2b receptor, Adenosine A2bR, adenosine receptor A2b, ADORA2, ADORA2B | |
| Human Adenosine A2b R synthetic peptide, Accession # NP_000667 | |
| 100 μg | |
| Primary | |
| Reconstitution default not known | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG |
| Flow Cytometry | |
| 2683F | |
| Flow Cytometry 0.25-1 ug/106 cells | |
| NP_000667 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 136 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction