missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ADCY10 (aa 1495-1581) Control Fragment Recombinant Protein

Product Code. 30212871
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212871

Brand: Invitrogen™ RP90979

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53290 (PA5-53290. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SAC belongs to a distinct class of mammalian adenylyl cyclase that is soluble and insensitive to G protein or forskolin regulation. It is localized in the cytoplasm and is thought to function as a general bicarbonate sensor throughout the body. It may also play an important role in the generation of cAMP in spermatozoa, implying possible roles in sperm maturation through the epididymis, capacitation, hypermotility, and/or the acrosome reaction.The protein encoded by this gene belongs to a distinct class of mammalian adenylyl cyclase that is soluble and insensitive to G protein or forskolin regulation. It is localized in the cytoplasm and is thought to function as a general bicarbonate sensor throughout the body. It may also play an important role in the generation of cAMP in spermatozoa, implying possible roles in sperm maturation through the epididymis, capacitation, hypermotility, and/or the acrosome reaction. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96PN6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55811
Name Human ADCY10 (aa 1495-1581) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 3',5'-cyclic AMP synthetase; 4930431D04Rik; 4931412F17; Adcy10; adenylate cyclase 10; adenylate cyclase 10 (soluble); adenylate cyclase 10, soluble; Adenylate cyclase homolog; Adenylate cyclase type 10; AH-related protein; ATP pyrophosphate-lyase; epididymis secretory sperm binding protein Li 7 A; germ cell soluble adenylyl cyclase; HCA2; HEL-S-7 A; hsAC; RP1-313L4.2; Sac; SACI; Sacy; soluble adenylyl cyclase; testicular soluble adenylyl cyclase; testicular soluble adenylyl cyclase (SAC)
Common Name ADCY10
Gene Symbol ADCY10
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NLENLVAQNTTGPVFCPRLYHLMAYVCILMGDGQKCGLFLNTALRLSETQGNILEKCWLNMNKESWYSTSELKEDQWLQTILSLPSW
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.