missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAMTS5 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2198
Additional Details : Weight : 0.00970kg
Description
ADAMTS5 Monoclonal specifically detects ADAMTS5 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ADAMTS5 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Q9UNA0 | |
ADAMTS5 | |
Mouse myeloma cell line NS0-derived recombinant human ADAMTS5 Ser262-Pro622 Accession # Q9UNA0 | |
100 μg | |
Primary | |
Detects human ADAMTS5 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) ADAMTS1 or rhADAMTSL1.2 is observed. | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
362810 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
A disintegrin and metalloproteinase with thrombospondin motifs 11, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 5 (aggrecanase-2), ADAM metallopeptidase with thrombospondin type 1 motif, 5, ADAM-TS 11, ADAM-TS 5, ADAMTS11, ADAMTS-11, ADAM-TS5, ADAMTS-5, ADMP2, ADMP-2, ADMP-2A disintegrin and metalloproteinase with thrombospondin motifs 5, Aggrecanase 2, Aggrecanase-2, disintegrin-like and metalloprotease with thrombospondin type 1 motif, 510ADAMTS11FLJ36738, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.14, EC 3.4.24.82 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
11096 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
For Research Use Only