missing translation for 'onlineSavingsMsg'
Learn More

Human ADAMTS4 Antibody, R&D Systems™

Mouse Monoclonal Antibody

£158.00 - £524.00

Specifications

Antigen ADAMTS4
Clone 416608
Dilution ELISA
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18721017
View Documents
R&D Systems
MAB43071-SP
25 μg
£158.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18713577
View Documents
R&D Systems
MAB43071-100
100 μg
£524.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ADAMTS4 Monoclonal antibody specifically detects ADAMTS4 in Human samples. It is validated for ELISA
Specifications

Specifications

ADAMTS4
ELISA
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 4, ADAM metallopeptidase with thrombospondin type 1 motif, 4, ADAM-TS 4, ADAMTS-2, ADAM-TS4, ADAMTS-4, ADMP-1, ADMP-1EC 3.4.24.82, Aggrecanase 1, aggrecanase-1, EC 3.4.24, KIAA0688A disintegrin and metalloproteinase with thrombospondin motifs 4
Mouse myeloma cell line NS0-derived recombinant human ADAMTS4, Phe213-Cys685, Accession # O75173
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
416608
ELISA
Unconjugated
Mouse
Human
O75173
9507
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.