missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAMTS15 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB5149U-100UG
This item is not returnable.
View return policy
Description
ADAMTS15 Monoclonal specifically detects ADAMTS15 in Human samples. It is validated for Flow Cytometry.
Specifications
| ADAMTS15 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8TE58 | |
| ADAMTS15 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ADAMTS15 Gly18-Cys682 Accession # Q8TE58 | |
| RUO | |
| 170689 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 561819 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| A disintegrin and metalloproteinase with thrombospondin motifs 15, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 15, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 15, preproprotein, ADAM metallopeptidase with thrombospondin type 1 motif, 15, ADAM-TS 15, ADAM-TS15, ADAMTS-15, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.14, EC 3.4.24.82, metalloprotease disintegrin 15 with thrombospondin domains, MGC126403 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human ADAMTS15 in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction