missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM32 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF5380
This item is not returnable.
View return policy
Description
ADAM32 Polyclonal specifically detects ADAM32 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ADAM32 | |
| Polyclonal | |
| Unconjugated | |
| Q8TC27 | |
| ADAM32 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ADAM32 Ser17-Thr476 Accession # Q8TC27 | |
| 100 μg | |
| Primary | |
| Detects human ADAM32 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) ADAM9, rhADAM22, and rhADAM23 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| a disintegrin and metalloprotease domain 32, a disintegrin and metalloproteinase domain 32, ADAM 32, ADAM metallopeptidase domain 32, disintegrin and metalloproteinase domain-containing protein 32, FLJ26299, FLJ29004, metalloproteinase 12-like protein | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 203102 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction