missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ADAM30 (aa 494-613) Control Fragment Recombinant Protein

Product Code. 30212484
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30212484 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30212484 Supplier Invitrogen™ Supplier No. RP89261

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (61%), Rat (61%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55288 (PA5-55288. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene is testis-specific and contains a polymorphic region, resulting in isoforms with varying numbers of C-terminal repeats.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UKF2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 11085
Name Human ADAM30 (aa 494-613) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 30; ADAM; ADAM 30; ADAM metallopeptidase domain 30; ADAM30; ADAMs; Disintegrin and metalloproteinase domain-containing protein 30; metalloendopeptidases; svph4; UNQ2509/PRO5997
Common Name ADAM30
Gene Symbol ADAM30
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RCFRKGCRSRYMQCQSIFGPDAMEAPSECYDAVNLIGDQFGNCEITGIRNFKKCESANSICGRLQCINVETIPDLPEHTTIISTHLQAENLMCWGTGYHLSMKPMGIPDLGMINDGTSCG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.