missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM22 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB49081
This item is not returnable.
View return policy
Description
ADAM22 Monoclonal specifically detects ADAM22 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ADAM22 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q9P0K1 | |
| ADAM22 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ADAM22 Gly26-His528 Accession # Q9P0K1 | |
| 100 μg | |
| Primary | |
| Detects human ADAM22 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human ADAM8, 9, 10, 12, 15, 19, 23, 32, or 33 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 498805 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ADAM 22, ADAM metallopeptidase domain 22, MDC2, metalloproteinase-disintegrin ADAM22-3, metalloproteinase-like, disintegrin-like, and cysteine-rich protein 2, MGC149832 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 53616 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction