missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM15 Ectodomain Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC935R-100UG
This item is not returnable.
View return policy
Description
ADAM15 Monoclonal specifically detects ADAM15 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| ADAM15 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q13444 | |
| ADAM15 | |
| COS-7 African green monkey SV40 transformed kidney fibroblast-like cell line-derived recombinant human ADAM15 Asp207-Thr696 Accession # Q13444 | |
| RUO | |
| 8751 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 23G9 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC15, MDC-15, Metargidin | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects the ectodomain of recombinant human (rh) ADAM15 in direct ELISAs and Western blots. In direct ELISAs, shows 100% cross-reactivity with recombinant mouse (rm) ADAM15 but does not cross-react with rhADAM8, 9, 17, 28, rmADAM9, or 10. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction