missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB14271-SP
This item is not returnable.
View return policy
Description
ADAM10 Monoclonal specifically detects ADAM10 in Human samples. It is validated for ELISA.
Specifications
| ADAM10 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian, Kuzbanian protein homolog, MADM, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease | |
| Spodoptera frugiperda, Sf 21 (baculovirus)-derived human ADAM10 protein, Thr214-Glu672, with a C-terminal 10-His tag, Accession No. O14672 | |
| 25 μg | |
| 102.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 172023 | |
| ELISA | |
| O14672 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human ADAM10 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction