missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ADA ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Human ADA quantitates human ADA in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ADA.
Catalyzes the hydrolytic deamination of adenosine and 2-deoxyadenosine. Plays an important role in purine metabolism and in adenosine homeostasis. Modulates signaling by extracellular adenosine, and so contributes indirectly to cellular signaling events. May act as a positive regulator of T-cell coactivation.
Specifications
Specifications
| Accession Number | Ada; ada1; Adenosine aminohydrolase; adenosine deaminase; RP11-61L14.5; zgc:92028 |
| Assay Range | 21 to 5,000 pg/mL |
| Assay Sensitivity | 21 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 100 |
| Gene Symbol | ADA |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction