missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ACVR1 Synthetic Peptide

Product Code. 15338940
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 15338940

Brand: Invitrogen™ PEP0724

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

ACVR1 Synthetic Peptide, PEP-0724, from Invitrogen.

Activin A receptor, type I (ACVR1) belongs to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. ACVR1 signals a specific transcriptional response in concert with activin type II receptors. The ACVR1 gene encodes activin A type I receptor, which signals a particular transcriptional response in concert with activin type II receptors. Mutations in ACVR1 have been linked to Fibrodysplasia Ossificans Progressiva (FOP), a rare genetic disorder characterized by the formation of bone in muscles, tendons, and other connective tissues, as well as Epicanthus.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 90
Species Human
Purification Method purified
Accession Number Q04771
Common Name Human ACVR1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol ACVR1
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.