missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Activin RIIA APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB340A
This item is not returnable.
View return policy
Description
Activin RIIA Monoclonal specifically detects Activin RIIA in Human samples. It is validated for Flow Cytometry.
Specifications
| Activin RIIA | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| activin A receptor, type II, activin A receptor, type IIA, Activin receptor type IIA, activin receptor type-2A, ActivinRIIA, ACTRII, ACTRIIA, ACVR2A, ACVR2ACTR-IIA, AVR2A, EC 2.7.11, EC 2.7.11.30 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 92 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 126706 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P27037 | |
| ACVR2A | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Activin RIIA Ala20-Pro134 Accession # P27037 | |
| 100 Tests | |
| Primary | |
| Detects human Activin RIIA in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) Activin RIA, rhActivin RIB, or rhActivin RIIB. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction