missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Activin RIB/ALK-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2221
This item is not returnable.
View return policy
Description
Activin RIB/ALK-4 Monoclonal specifically detects Activin RIB/ALK-4 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Activin RIB/ALK-4 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P36896 | |
| ACVR1B | |
| Mouse myeloma cell line NS0-derived recombinant human Activin RIB/ALK-4 Ser24-Glu126 Accession # P36896 | |
| 100 μg | |
| Primary | |
| Detects human Activin RIB/ALK-4 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Activin RIA, RIIA, or RIIB is observed. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 544310 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| activin A receptor, type IB, Activin receptor type IB, Activin receptor-like kinase 4, ActivinRIB, ActRIB, ACVR1B, ACVRLK4ACTR-IB, ALK4, ALK-4, ALK4activin A receptor, type II-like kinase 4, EC 2.7.11, EC 2.7.11.30, Serine/threonine-protein kinase receptor R2, SKR2activin receptor type-1B | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 91 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction