missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Activin A Recombinant Protein

Product Code. p-7121650
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11300862

Brand: Gibco™ PHC9561

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute lyophilized recombinant human Activin A in sterile PBS containing 0.1% human serum albumin. Further dilutions should be made in low endotoxin medium or a buffered solution containing a carrier protein such as heat inactivated FCS or tissue culture grade BSA. Storage: Store this lyophilized preparation at = -20°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at = -20°C. Avoid repeated freeze/thaw cycles.

Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I ( I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Activin A mutations are associated with fibrodysplasia ossificans progressive.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08476, Q04998
For Use With (Application) Bioactivity
Formulation PBS with no preservative
Gene ID (Entrez) 16323, 3624
Molecular Weight (g/mol) 25 kDa
Name Human Activin A
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/mL
Gene Alias activin A; activin beta-A chain; BMP7; BMP-7; Bone morphogenetic protein 7; EDF; Eptotermin alfa; erythroid differentiation factor; erythroid differentiation protein; follicle-stimulating hormone-releasing protein; FRP; FSH-releasing protein; INHB; Inhba; inhibin B subunit; inhibin beta A chain; inhibin beta A protein; inhibin beta A subunit; inhibin beta-A; inhibin beta-A-subunit; inhibin beta-subunit; inhibin subunit beta A; inhibin, beta A; inhibin, beta A (activin A, activin AB alpha polypeptide); Inhibin, beta-1; OP1; OP-1; Osteogenic protein 1; unnamed protein product
Common Name Activin A
Gene Symbol Inhba
Biological Activity ED50 = 0.5 - 5 ng/mL; determined by the dose-dependent proliferation of MPC-11 cells.
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Content And Storage -20°C
Classification Carrier-Free
Activity ED50 = 0.5 - 5 ng/mL; determined by the dose-dependent proliferation of MPC-11 cells.
Bioassay Inhibition of Proliferation of MPC11 cells
Endotoxin Level <1 EU/ml
Shipping Condition Dry Ice
Expression System HEK293 cells
Protein Family Growth Factors & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype Activins
Research Category Neurobiology, Stem Cell Research, Differentiation
Product Line Gibco
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.