missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Actin-like 7B (aa 229-307) Control Fragment Recombinant Protein

Product Code. 30200230
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200230

Brand: Invitrogen™ RP93002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55147 (PA5-55147. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a member of a family of actin-related proteins which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene, and related gene, ACTL7A, are intronless, and are located approximately 4 kb apart in a head-to-head orientation within the familial dysautonomia candidate region on 9q31. Based on mutational analysis of the ACTL7B gene in patients with this disorder, it was concluded that it is unlikely to be involved in the pathogenesis of dysautonomia. Unlike ACTL7A, the ACTL7B gene is expressed predominantly in the testis, however, its exact function is not known.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y614
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10880
Name Human Actin-like 7 B (aa 229-307) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias actin like 7 B; actin-like 7 B; actin-like 7-beta; actin-like protein 7 B; Actin-like-7-beta; ACTL7B; MGC151192; MGC151194; RP11-3J11.2; Tact1; t-actin 1; T-actin-1; testis tissue sperm-binding protein Li 43 A; testis-specific actin-1; Unknown (protein for MGC:134108)
Common Name Actin-like 7 B
Gene Symbol ACTL7B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LTNYLMQLLNEAGHAFTDDHLHIIEHIKKKCCYAAFLPEEELGLVPEELRVDYELPDGKLITIGQERFRCSEMLFQPSL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.