missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ACP6 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7766-SP
This item is not returnable.
View return policy
Description
ACP6 Polyclonal specifically detects ACP6 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ACP6 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| acid phosphatase 6, lysophosphatidicPACPL1, Acid phosphatase-like protein 1, ACPL1, ACPL1acid phosphatase like 1, EC 3.1.3.2, LPAP, LPAPlysophosphatidic acid phosphatase type 6, PACPL1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 51205 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| BAA91310 | |
| ACP6 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ACP6 Glu33-Glu428 Accession # BAA91310 | |
| 25 μg | |
| Primary | |
| Detects human ACP6 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction