missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ACLP Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC6425R-100UG
This item is not returnable.
View return policy
Description
AE Binding Protein 1/ACLP Monoclonal specifically detects AE Binding Protein 1/ACLP in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| AE Binding Protein 1/ACLP | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8IUX7 | |
| AEBP1 | |
| Mouse myeloma cell line NS0-derived recombinant human ACLP Glu345-Phe1158 Accession # Q8IUX7 | |
| RUO | |
| 165 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 590831 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ACLP, ACLPadipocyte enhancer binding protein 1, AE binding protein 1, AE-binding protein 1adipocyte enhancer-binding protein 1, AEBP1, Aortic carboxypeptidase-like protein, FLJ33612 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human ACLP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse ACLP is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction