missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Acinus (CP) Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Acinus (CP) Synthetic Peptide, PEP-0063, from Invitrogen.
ACINUS (apoptotic chromatin condensation inducer in the nucleus) is an induced of chromatin condensation. ACINUS is cleaved by caspase-3 and an additional unknown protease generating a small active peptide, p17, which causes chromatin condensation in vitro when it is added to purified nuclei. ACINUS also induces apoptotic chromatin condensation in cells. ACINUS is ubiquitously expressed. Three different spliced forms of Acinus have been identified in human and mouse.
Specifications
Specifications
| Gene ID (Entrez) | 22985 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q9UKV3 |
| Common Name | Human Acinus (CP) |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction