missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Acetylcholinesterase/ACHE Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7574
This item is not returnable.
View return policy
Description
Acetylcholinesterase/ACHE Monoclonal specifically detects Acetylcholinesterase/ACHE in Human samples. It is validated for Immunohistochemistry.
Specifications
| Acetylcholinesterase/ACHE | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 43 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 872044 | |
| Immunohistochemistry 8-25 ug/mL | |
| P22303 | |
| ACHE | |
| Chinese hamster ovary cell line CHO-derived recombinant human ACHE Met1-Leu614 Accession # P22303 | |
| 100 μg | |
| Primary | |
| Detects human ACHE in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction